Merck
  • Solid-phase microextraction-capillary gas chromatography combined with microwave-induced plasma atomic-emission spectrometry for selenite determination.
Solid-phase microextraction-capillary gas chromatography combined with microwave-induced plasma atomic-emission spectrometry for selenite determination.
Analytical and bioanalytical chemistry (2004-06-29)
Emmanouil Dimitrakakis, Christina Haberhauer-Troyer, Yo Abe, Maria Ochsenkühn-Petropoulou, Erwin Rosenberg
ABSTRACT
The use of solid-phase microextraction (SPME) with gas chromatography coupled to microwave-induced plasma atomic-emission detection (GC-MIP-AED) is described for selenite [Se(IV)] speciation. Aqueous standards were derivatised with sodium tetraethyl- or tetrapropylborate and extracted by SPME. Headspace extraction of the ethyl and propyl derivatives was studied. Relevant experimental conditions were optimised, including conditions for derivatisation and extraction and those of gas chromatographic analysis. The limits of detection achieved for headspace sampling of derivatised Se(IV) were in the low ng mL(-1) range for both ethylation and propylation. When the method was applied to analysis of selenite in selenised yeast reference material results were in good agreement with the indicated values.
Product Description
Supelco
SPB®-Octyl Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 1.00 μm
Supelco
SPB®-Octyl Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.25 μm