Merck
  • The gap between scientific evidence and clinical practice: 5-aminosalicylates are frequently used for the treatment of Crohn's disease.
The gap between scientific evidence and clinical practice: 5-aminosalicylates are frequently used for the treatment of Crohn's disease.
Alimentary pharmacology & therapeutics (2014-08-26)
A M Schoepfer, M Bortolotti, V Pittet, C Mottet, J-J Gonvers, O Reich, N Fournier, J-P Vader, B Burnand, P Michetti, F Froehlich
ABSTRACT
There is uncertain evidence of effectiveness of 5-aminosalicylates (5-ASA) to induce and maintain response and remission of active Crohn's disease (CD), and weak evidence to support their use in post-operative CD. To assess the frequency and determinants of 5-ASA use in CD patients and to evaluate the physicians' perception of clinical response and side effects to 5-ASA. Data from the Swiss Inflammatory Bowel Disease Cohort, which collects data since 2006 on a large sample of IBD patients, were analysed. Information from questionnaires regarding utilisation of treatments and perception of response to 5-ASA were evaluated. Logistic regression modelling was performed to identify factors associated with 5-ASA use. Of 1420 CD patients, 835 (59%) were ever treated with 5-ASA from diagnosis to latest follow-up. Disease duration >10 years and colonic location were both significantly associated with 5-ASA use. 5-ASA treatment was judged to be successful in 46% (378/825) of treatment episodes (physician global assessment). Side effects prompting stop of therapy were found in 12% (98/825) episodes in which 5-ASA had been stopped. 5-Aminosalicylates were frequently prescribed in patients with Crohn's disease in the Swiss IBD cohort. This observation stands in contrast to the scientific evidence demonstrating a very limited role of 5-ASA compounds in the treatment of Crohn's disease.
Product Description
Supelco
Mesalamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
5-Aminosalicylic acid, 95%
Sigma-Aldrich
5-Aminosalicylic acid, ≥99%
Mesalazine, European Pharmacopoeia (EP) Reference Standard
Mesalazine for system suitability, European Pharmacopoeia (EP) Reference Standard