Merck
  • Modified Fenton oxidation of diesel fuel in arctic soils rich in organic matter and iron.
Modified Fenton oxidation of diesel fuel in arctic soils rich in organic matter and iron.
Chemosphere (2014-07-30)
Mary K Sherwood, Daniel P Cassidy
ABSTRACT
Modified Fenton (MF) chemistry was tested in the laboratory to treat three diesel fuel-contaminated soils from the Canadian arctic rich in soil organic matter (SOM) and Fe oxides. Reactors were dosed with hydrogen peroxide (HP), and treatment was compared in reactors with SOM as the only chelate vs. reactors to which ethylenediaminetetraacetate (EDTA) was added. Concentrations of diesel fuel and HP were measured over time, and the oxidation of both diesel fuel and SOM were quantified in each soil. A distinct selectivity for oxidation of diesel fuel over SOM was observed. Reactors with EDTA showed significantly less diesel fuel oxidation and lower oxidant efficiency (diesel fuel oxidized/HP consumed) than reactors with SOM as the only chelate. The results from these studies demonstrate that MF chemistry can be an effective remedial tool for contaminated arctic soils, and challenge the traditional conceptual model that SOM reduces the efficiency of MF treatment through excessive scavenging of oxidant.
Product Description
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Iron(III) oxide, dispersion, nanoparticles, ≤110 nm particle size, 15 wt. % in ethanol
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Iron, IRMM®, certified reference material, 0.5 mm wire
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, 98.5-101.5%, BioUltra
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid calcium disodium salt hydrate
USP
Edetate disodium, United States Pharmacopeia (USP) Reference Standard
Iron, foil, not light tested, 100x100mm, thickness 0.008mm, 99.85%
Iron, foil, light tested, 50x50mm, thickness 0.05mm, hard, 99.5%
Iron, foil, not light tested, 25x25mm, thickness 0.009mm, 99.85%
Iron, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Iron, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%
Iron, foil, 8mm disks, thickness 0.20mm, hard, 99.5%
Iron, foil, not light tested, 25x25mm, thickness 0.002mm, 99.85%
Iron, foil, 8mm disks, thickness 0.38mm, hard, 99.5%
Iron, foil, not light tested, 300x300mm, thickness 0.05mm, hard, 99.5%
Iron, rod, 300mm, diameter 5.0mm, as drawn, 99.99+%
Iron, foil, light tested, 25x25mm, thickness 0.025mm, hard, 99.5%