Merck
Development of serum zinc as a biological dosimeter in mice.
International journal of radiation biology (2014-05-16)
Xuan-Yu Min, Xiao-Hong Zhang, Qi-Pu Zhou, Xiao-Dan Hu, Pei-Dang Liu, Hai-Qian Zhang
ABSTRACT
To develop a new biological dosimeter based on serum zinc concentration. Male mice (8 weeks old) were exposed to different doses (0, 1.0, 2.0, 4.0, or 8.0 Gy) of gamma rays from a (60)Co source. Blood was then collected from the orbital area of these mice, and the serum zinc concentration was detected using the 2-(5-bromo-2-pyridylazo)-5-diethylaminophenol colorimetric method. The data were analyzed using one-way analysis of variance. The serum zinc concentration in the irradiated mice decreased with increasing dose. Two dose-response relationships fitted to the linear quadratic curve were obtained: One immediately after exposure (y = 0.010x(2) - 0.133x + 0.663, r = 0.983) and the other on the seventh day after exposure (y = 0.008x(2) - 0.127x + 0.695, r = 0.990). The serum zinc concentration continued to decrease until 21 days after exposure. The absorbed doses estimated using both dose-response relationships were close to the actual doses. Serum zinc is a quick, effective, and sensitive biomarker for early biological doses assessment of mice irradiated by gamma radiation.
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Zinc, wire reel, 10m, diameter 0.5mm, as drawn, 99.99+%
Zinc, wire reel, 5m, diameter 0.05mm, as drawn, 99%
Zinc, wire reel, 0.1m, diameter 6.35mm, 99.99+%
Zinc, wire reel, 1m, diameter 1.0mm, 99.99+%
Zinc, wire reel, 5m, diameter 0.025mm, as drawn, 99%
Zinc, wire reel, 100m, diameter 2.0mm, extruded, 99.9%
Zinc, wire reel, 25m, diameter 0.125mm, as drawn, 99%
Zinc, wire reel, 20m, diameter 2.0mm, extruded, 99.9%
Zinc, wire reel, 10m, diameter 0.25mm, as drawn, 99.99+%
Zinc, wire reel, 2m, diameter 1.0mm, 99.999%
Zinc, foil, 8mm disks, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 0.015mm, 99.9%
Zinc, foil, not light tested, 150x150mm, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, light tested, 150x150mm, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 5m coil, thickness 0.25mm, as rolled, 99.95+%
Zinc, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 8mm disks, thickness 0.35mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 1.0mm, as rolled, 99.99+%