Merck
Decompositional odor analysis database.
Journal of forensic sciences (2004-08-20)
Arpad A Vass, Rob R Smith, Cyril V Thompson, Michael N Burnett, Dennis A Wolf, Jennifer A Synstelien, Nishan Dulgerian, Brian A Eckenrode
ABSTRACT
This study, conducted at the University of Tennessee's Anthropological Research Facility (ARF), describes the establishment of the Decompositional Odor Analysis (DOA) Database for the purpose of developing a man-portable, chemical sensor capable of detecting clandestine burial sites of human remains, thereby mimicking canine olfaction. This "living" database currently spans the first year and a half of burial, providing identification, chemical trends and semi-quantitation of chemicals liberated below, above and at the surface of graves 1.5 to 3.5 ft deep (0.45 to 1.0 m) for four individuals. Triple sorbent traps (TSTs) were used to collect air samples in the field and revealed eight major classes of chemicals containing 424 specific volatile compounds associated with burial decomposition. This research is the first step toward identification of an "odor signature" unique to human decomposition with projected ramifications on cadaver dog training procedures and in the development of field portable analytical instruments which can be used to locate human remains buried in shallow graves.
Product Description
Swagelok® Reducer, stainless steel, 1/4 in. Swagelok, for 1/8 in. tube
Swagelok® Reducer, Swagelok® 400-R-2, brass, 1/4 in. Swagelok, for 1/8 in. tube