Merck
  • Tin-catalyzed conversion of biomass-derived triose sugar and formaldehyde to α-hydroxy-γ-butyrolactone.
Tin-catalyzed conversion of biomass-derived triose sugar and formaldehyde to α-hydroxy-γ-butyrolactone.
Chemical communications (Cambridge, England) (2014-03-29)
Sho Yamaguchi, Ken Motokura, Yasuharu Sakamoto, Akimitsu Miyaji, Toshihide Baba
ABSTRACT
The direct conversion of biomass-derived 1,3-dihydroxyacetone (DHA) and formaldehyde to α-hydroxy-γ-butyrolactone (HBL) was achieved through the use of tin(iv) chloride and a small amount of water and the yield reached up to 70%. The reaction mechanism was also investigated by incorporating d2-formaldehyde into the reaction mixtures.
Sigma-Aldrich
α-Hydroxy-γ-butyrolactone, technical grade
Pricing and availability is not currently available.