Merck
  • Synthesis of γ- and δ-lactone natural products by employing a trans-cis isomerization/lactonization strategy.
Synthesis of γ- and δ-lactone natural products by employing a trans-cis isomerization/lactonization strategy.
Chemical & pharmaceutical bulletin (2013-04-03)
Machiko Ono, Keisuke Kato, Hiroyuki Akita
ABSTRACT
Alkaline hydrolysis of 4-hydroxy- or/and 5-hydroxy-(2E)-alkenoate followed by acid treatment gave the corresponding (2E)-alkenoic acids which were subjected to lactone formation reaction without further purification. The crude acids were treated with 2,4,6-trichlorobenzoyl chloride in pyridine to afford γ-lactone or δ-lactone, respectively, accompanied by trans-cis isomerization. By this procedure, (±)-(4,5)-trans-5-benzyloxy-2-hexen-4-olide (90% overall yield), (S)-5-hydroxy-2-penten-4-olide (86% overall yield), (4S,5R)-5-hydroxy-2-hexen-4-olide (86% overall yield), (4R,5S)-5-hydroxy-2-hexen-4-olide (82% overall yield), (S)-parasorbic acid (58% overall yield) and natural product, (5R,7S)-7-hydroxy-2-octen-5-olide (euscapholide: 20% overall yield) were synthesized.
Sigma-Aldrich
2,4,6-Trichlorobenzoyl chloride, 97%