Merck
  • Comparison of exposure assessment methods in occupational exposure to benzene in gasoline filling-station attendants.
Comparison of exposure assessment methods in occupational exposure to benzene in gasoline filling-station attendants.
Toxicology letters (2005-11-18)
Mariella Carrieri, Enrico Bonfiglio, Maria Luisa Scapellato, Isabella Maccà, Giovanna Tranfo, Paolo Faranda, Enrico Paci, Giovanni Battista Bartolucci
ABSTRACT
The aim of this study was to assess gasoline filling-station attendants' exposure to benzene and to determine which biological exposure index (BEI), trans,trans-muconic acid (t,t-MA) or S-phenylmercapturic acid (S-PMA), shows better correlation with environmental exposure. Exposure to benzene was measured using passive samplers (Radiello) attached to the collar of the overalls of subjects (n=33) just before the work-shift (approximately 8h); analysis was performed by GC-FID. S-PMA and t,t-MA were determined, respectively, by an immunochemiluminescent assay based on specific monoclonal antibodies and by HPLC-UV at 264 nm. Both methods of biological monitoring were performed on beginning and end-shift urine samples, and expected t,t-MA and S-PMA values were calculated. Smoking habits and life-style were ascertained by means of a questionnaire. Both environmental and biological monitoring data showed that benzene exposure for gasoline filling-station attendants was low when compared with the respective ACGIH limit values (means-benzene: 0.044 mg/m(3); t,t-MA: 171 microg/g creatinine; S-PMA: 2.7 microg/g creatinine). No significant correlation was found between exposure to benzene and t,t-MA or S-PMA excretion data. The use of expected values was also experimented for S-PMA and t,t-MA. This consists of calculating, on the basis of the known half-life of the benzene metabolite, the concentration of that metabolite that a worker should present at the end of the work-shift, the difference between this value and the value actually found is a measure of benzene exposure during work. The use of expected values in biological monitoring did not improve correlations. At these low benzene levels, environmental monitoring seems to be the best method of evaluating individual exposure. However, biological monitoring remains useful, as a mean of assessing group exposure.
Product Description
radiello BTEX/VOC Starter Kit, Thermal Desorption
radiello Empty Storage Tubes, polypropylene (with stopper), volume 12 mL, pk of 100
Supelco
radiello BTEX Calibration Kit (Thermal Desorption)
radiello Clips, suspending Radiello triangular support plate, pk of 20
radiello Filtration Kit, pk of 20
radiello On-Field Thermometer, Includes one standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use, pk of 3
radiello BTEX/VOC Starter Kit, CS2 Desorption
radiello Diffusive Bodies, white, configured for general use, pk of 20
radiello On-Field Thermometer, Reader, pk of 1
radiello Diffusive Bodies, blue, configured for sampling light sensitive compounds, pk of 20
Supelco
radiello BTEX Calibration Kit (CS2 Desorption)
radiello Empty Storage Tubes, glass (with stopper), volume 2.8 mL, pk of 100
radiello Diffusive Bodies, yellow, configured for reduced sampling rates, pk of 20
radiello Diffusive Bodies, permeative, configured for anaesthetic gases and vapors, pk of 20
Supelco
radiello Aldehyde Calibration Standard, 50 μg/mL each component in acetonitrile, pkg of 10 mL
radiello 1,3-Butadiene Starter Kit, Thermal Desorption, pkg of 1 kit (2 RAD141 adsorbing cartridges and barcode labels, 1 RAD1202 yellow diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet)
Supelco
radiello Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide), pkg of 100 mL
radiello Hydrogen Sulfide Starter Kit
radiello Barcode Labels, pk of 198
radiello Anesthetic Gases and Vapor Sampler
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (CS2 Desorption), matrix SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh), pk of 20
radiello Cartridge Adsorbents, for sampling Ammonia (NH3), matrix microporous PE impregnated with phosphoric acid, pk of 20
radiello Outdoor Shelter, pk of 10
radiello Replacement Mounting Strips, for use with Radiello Outdoor Shelter, pk of 100
radiello Cartridge Adsorbents, for sampling Aldehydes, matrix SS net with 2,4-DNPH coated FLORISIL®, pk of 20
radiello Cartridge Adsorbents, for sampling Anaesthetic Gases/Vapors, matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh), pk of 20
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (thermal desorption), matrix SS net (3 x 8 μm, 4.8 mm diam.), Carbograph, pk of 20
radiello Triangular Support Plate, pk of 20
radiello Vertical Adapter, threaded for standard use, pk of 20