Merck
  • Oral bullous eruption after taking lisinopril--case report and literature review.
Oral bullous eruption after taking lisinopril--case report and literature review.
Wiener klinische Wochenschrift (2013-06-25)
Marinka Baričević, Marinka Mravak Stipetić, Mirna Situm, Branka Marinović, Sven Seiwerth, Denis Baričević, Božana Lončar
ABSTRACT
Drug-induced lesions of oral mucosa are well-established side effect of different commonly used drugs. A female patient under treatment for hypertension with an angiotensin-converting enzyme inhibitor (ACE inhibitor), lisinopril, developed blisters and ulcerations on oral mucosa 3 weeks after lisinopril intake. Due to clinical finding drug-induced pemphigus was considered. However, direct and indirect immunofluorescence anal-ysis revealed no autoantibodies that are commonly present in pemphigus while histological study suggested allergic reaction. Lisinopril was discontinued from further therapy and after a month after her first arrival patient has experienced complete remission of the disease. This case raises the question, whether the term pemphigus in drug-induced reactions could be used when immunopathological criteria for pemphigus are not fulfilled.
Sigma-Aldrich
Lisinopril, ≥98% (HPLC)
Supelco
Lisinopril, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Lisinopril, United States Pharmacopeia (USP) Reference Standard
Lisinopril dihydrate, European Pharmacopoeia (EP) Reference Standard