Merck
Primer on agar-based microbial imaging mass spectrometry.
Journal of bacteriology (2012-07-24)
Jane Y Yang, Vanessa V Phelan, Ryan Simkovsky, Jeramie D Watrous, Rachelle M Trial, Tinya C Fleming, Roland Wenter, Bradley S Moore, Susan S Golden, Kit Pogliano, Pieter C Dorrestein
ABSTRACT
Matrix-assisted laser desorption ionization-time of flight (MALDI-TOF) imaging mass spectrometry (IMS) applied directly to microbes on agar-based medium captures global information about microbial molecules, allowing for direct correlation of chemotypes to phenotypes. This tool was developed to investigate metabolic exchange factors of intraspecies, interspecies, and polymicrobial interactions. Based on our experience of the thousands of images we have generated in the laboratory, we present five steps of microbial IMS: culturing, matrix application, dehydration of the sample, data acquisition, and data analysis/interpretation. We also address the common challenges encountered during sample preparation, matrix selection and application, and sample adherence to the MALDI target plate. With the practical guidelines described herein, microbial IMS use can be extended to bio-based agricultural, biofuel, diagnostic, and therapeutic discovery applications.
Product Description
Millipore
Agar, suitable for microbiology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Noble agar
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Sigma-Aldrich
Select agar
Sigma-Aldrich
Agar, suitable for plant cell culture
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Agar, ash 2.0-4.5%
Millipore
Agar, powder, suitable for microbiology
Millipore
Agar, high purity, low ionic content, low gel strenght, suitable for microbiology
Sigma-Aldrich
Bacteriological agar, for molecular biology