Merck
  • Polynuclear ruthenium, osmium and gold complexes. The quest for innovative anticancer chemotherapeutics.
Polynuclear ruthenium, osmium and gold complexes. The quest for innovative anticancer chemotherapeutics.
Current topics in medicinal chemistry (2011-11-02)
Christian G Hartinger, Andrew D Phillips, Alexey A Nazarov
ABSTRACT
Polynuclear compounds are a relatively new and successful approach in metal-based cancer chemotherapy as typified by the trinuclear Pt compound BBR3464 which was evaluated in clinical trials. In this review, we discuss newer developments of polynuclear ruthenium, osmium and gold complexes, focusing on their anticancer activity. The compounds presented are often supposed to exert their anticancer activity by different modes of action as compared to established drugs, including newly proposed mechanisms such as enzyme inhibition, crosslinking of biomacromolecules or through photo-activation, though many of the examples are also capable of binding to DNA nucleobases. Important metabolization and chemical characteristics of such compounds are discussed, and if the appropriate data is available, molecular modes of action are highlighted.
Sigma-Aldrich
Ruthenium black
Sigma-Aldrich
Osmium, powder, 99.9% trace metals basis
Sigma-Aldrich
Ruthenium, powder, −200 mesh, 99.9% trace metals basis
Ruthenium, Ruthenium, foil, 10x10mm, thickness 1.0mm, 99.9%
Sigma-Aldrich
Ruthenium, powder, 99.99% trace metals basis
Ruthenium, Ruthenium, foil, 6x6mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.025μm, specific density 30.5μg/cm2, permanent mylar 3.5μm support, 99.9%
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.1μm, specific density 122μg/cm2, permanent mylar 3.5μm support, 99.9%
Ruthenium, Ruthenium, pellets, 5g, max. size 10mm, 99.9%
Ruthenium, Ruthenium, pellets, 2.5g, max. size 10mm, 99.9%
Ruthenium, Ruthenium, rod, 12.7mm, diameter 12.7mm, 99.9%
Ruthenium, Ruthenium, bar, 25mm x 2mm x 2mm, 99.9%
Ruthenium, Ruthenium, bar, 50mm x 2mm x 2mm, 99.9%
Ruthenium, Ruthenium, foil, 25x25mm, thickness 1.0mm, 99.9%