Merck
  • GABA (γ-aminobutyric acid), a non-protein amino acid counters the β-adrenergic cascade-activated oncogenic signaling in pancreatic cancer: a review of experimental evidence.
GABA (γ-aminobutyric acid), a non-protein amino acid counters the β-adrenergic cascade-activated oncogenic signaling in pancreatic cancer: a review of experimental evidence.
Molecular nutrition & food research (2011-08-02)
Hussein A N Al-Wadei, Mohammad F Ullah, Mohammed Al-Wadei
ABSTRACT
GABA is a bioactive constituent of fruits, vegetables, cereals and is believed to play a role in defense against stress in plants. In animals, it acts as an inhibitory neurotransmitter in brain while also expressed in non-neuronal cells. Studies have implicated the regulator of fight or flight stress responses, β-AR signaling cascade, as mediators of cancer growth and progression in in vitro and in vivo models of pancreatic malignancies. Pancreatic cancer is the fourth leading cause of cancer mortality in western countries. This malignancy is generally unresponsive to conventional radio- and chemotherapy, resulting in mortality rate near 100% within 6 months of diagnosis. We review a series of experiments from our laboratory and those of others examining the contribution of this signaling network to pancreatic and other human malignancies. Stimulation of the β-adrenergic receptor by lifestyle and environmental factors, as well as a pre-existing risk of neoplasm, activates downstream effector molecules that lead to pro-oncogenic signaling and thereby aid cancer growth. GABAergic signaling mediated by the serpentine receptor GABA(B) acts as an antagonist to β-adrenergic cascade by intercepting adenylyl cyclase. These evidences enhance the pharmacological value of human diets rich in GABA for use as an adjuvant to standard therapies.
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Supelco
γ-Aminobutyric acid, analytical standard
Vigabatrin impurity D, European Pharmacopoeia (EP) Reference Standard