Merck
  • Is it safe to switch between efavirenz and nevirapine in the event of toxicity?
Is it safe to switch between efavirenz and nevirapine in the event of toxicity?
The Lancet. Infectious diseases (2007-10-27)
Ushma Mehta, Gary Maartens
ABSTRACT
The non-nucleoside reverse transcriptase inhibitors (NNRTIs) efavirenz and nevirapine are chemically distinct, but both may cause cutaneous hypersensitivity and hepatotoxicity. We reviewed the literature to assess the evidence for cross-reactivity between nevirapine and efavirenz. All papers, abstracts, or presentations, regardless of study design, that made reference to the response of patients who were switched from one NNRTI to another as a result of an adverse drug reaction were included. Most of the studies were retrospective. Recurrent reactions occurred in 30 (12.6%) of 239 reported patients with rash who were switched from nevirapine to efavirenz, compared with eight (50%) of 16 patients switched from efavirenz to nevirapine. Hepatitis did not recur in either the 11 reported patients switched from nevirapine to efavirenz, or in the single reported patient who was switched from efavirenz to nevirapine. Substituting efavirenz for nevirapine following hepatotoxicity or cutaneous hypersensitivity appears to be reasonable, providing that the adverse reaction to nevirapine was not life-threatening. There is insufficient evidence to recommend substituting nevirapine for efavirenz following either hepatotoxicity or cutaneous hypersensitivity.
Product Description
Nevirapine for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nevirapine
Nevirapine (anhydrous), European Pharmacopoeia (EP) Reference Standard