Merck
  • The anti-caries efficacy of calcium carbonate-based fluoride toothpastes.
The anti-caries efficacy of calcium carbonate-based fluoride toothpastes.
International dental journal (2005-07-12)
R J M Lynch, J M ten Cate
ABSTRACT
To summarise clinical support for the anti-caries efficacy of fluoride toothpastes containing sodium monofluorophosphate (SMFP) and to discuss the possible means by which the abrasive particles in calcium carbonate-based SMFP toothpastes might complement and/or enhance fluoride efficacy. The anti-caries efficacy of fluoride has been proven beyond any reasonable doubt, and the efficacy of SMFP, when incorporated into a variety of compatible toothpaste formulations, has been established in numerous clinical trials. Calcium carbonate-based toothpastes may also influence caries by effecting an increase in plaque calcium levels; an inverse relationship between plaque calcium and caries is well-established. It has also been reported that plaque fluoride levels are dependent on plaque calcium levels. Hence elevated plaque calcium resulting from the use of calcium carbonate-based toothpastes has the potential to elevate plaque fluoride, itself linked to reduced caries experience. It has been shown that calcium carbonate particles are retained by plaque and this may also influence caries by neutralising harmful plaque acids and concurrently liberating calcium. Fluoride delivered from calcium carbonate-based SMFP toothpastes is an effective means of reducing caries. Further, calcium carbonate may confer additional benefits through elevation of oral calcium levels and neutralisation of plaque-acids.
Product Description
USP
Calcium Carbonate (AS), United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
Supelco
Calcium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material