Merck
  • Update on nandrolone and norsteroids: how endogenous or xenobiotic are these substances?
Update on nandrolone and norsteroids: how endogenous or xenobiotic are these substances?
European journal of applied physiology (2004-03-26)
V Bricout, F Wright
ABSTRACT
Norsteroids are xenobiotics with androgenic and anabolic properties known since as far back as the 1930s. In doping controls, the use of the banned xenobiotic norsteroids is detected in the competitor's urines by the measurement of norandrosterone (19-NA) and noretiocholanolone (19-NE), which are the main metabolites for nandrolone (NT) and most norsteroids with anabolic properties. In 1996, the IOC subcommission "Doping and Biochemistry of Sport" informed the Heads of the "IOC Accredited Laboratories" that the recommended cut-off limit for reporting an offence was to be 1-2 ng ml(-1) urine for either 19-NA or 19-NE. We will discuss how technical progress in gas chromatography coupled to high-resolution mass spectrometry permitted a dramatic increase in sensitivity with a detection limit of 1 pg ml(-1) urine, or less, and an assay limit of 20-50 pg ml(-1) urine, for either 19-NA or 19-NE. As a paradox, norsteroids have been known for decades as not only xenobiotics but also obligatory endogenous intermediates in the biosynthesis of estrogens from androgens in all species, man included. It is this biochemical observation which fed the active scientific and medical controversy initiated in 1998 over the possibly endogenous production of nandrolone and metabolites well over the new IOC's recommended cut-off limit of 2 ng ml(-1) urine. Notwithstanding the particular technical difficulties attached, we will provide data and discuss the minute endogenous levels detected and measured in man either at rest, after performance or training and compare them to the relatively high levels reported in male athlete's doping controls today. We will also discuss data on the pharmacological effects of some contraceptive therapies containing norsteroids in women. In view of the well-documented noxious effects repeatedly observed after anabolic steroid misuse, the confirmation and implementation of technically proven procedures for reporting norsteroid abuse in sports seems an important enough goal to protect athlete's health against such abuses and justifies up dating the review of the patent scientific and medical experience and knowledge gained over the last 50 years on nandrolone and its minor production in man and woman.
Product Description
Supelco
Nandrolone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
19-Nortestosterone, ≥99.0% (HPLC)