- Hu 210: a potent tool for investigations of the cannabinoid system.
Hu 210: a potent tool for investigations of the cannabinoid system.
CNS drug reviews (2001-07-28)
A Ottani, D Giuliani
PMID11474421
ABSTRACT
The synthetic compound HU 210 displays a multiplicity of biochemical, pharmacological, and behavioral effects, most of which have been demonstrated to be dependent on a selective agonistic activity at CB(1) and CB(2) cannabinoid receptors and to involve the main neurotransmitter systems. Results obtained in various studies suggest a potential clinical application of this highly potent drug (e.g., as antipyretic, antiinflammatory, analgesic, antiemetic, and antipsychotic agent) as well as its usefulness in research aimed to develop a better understanding of the involvement of the endogenous cannabinoid system in a number of physiopathological functions.
Product Description
Supelco
HU-210 solution, 100 μg/mL in methanol, ampule of 1 mL, (Spice Cannabinoid), certified reference material, Cerilliant®