Merck
The role of busulfan in bone marrow transplantation.
Medical oncology (Northwood, London, England) (1999-10-19)
M Hassan
ABSTRACT
High-dose busulfan is an important component in many conditioning protocols for hematopoietic stem cell transplantation (HSCT) or bone marrow transplantation (BMT) in both adults and children. During the past 12y several studies have reported the wide inter-individual variability in busulfan disposition. Age, disease status, hepatic function, circadian rhythmicity, drug interactions and bioavailability, were identified as factors contributing to the high inter-individual variability found in busulfan disposition. Traditionally, a standard busulfan dose of 4mg/kg/d for four days is used in most BMT/HSCT protocols. Many investigations have pointed out the pharmacodynamic relationship between a high busulfan systemic exposure and the occurrence of BMT related toxicity including hepatic veno-occlusive disease (VOD), interstitial pneumonia and alopecia in adult patients. However, studies in young patients have shown a high rate of graft failure and subsequently relapse which most probably is due to the low systemic exposure despite the standard dose schedule. In children and infants VOD was not observed with the standard doses. Increasing interest for the drug and new modification strategies for children led to higher rate of VOD and CNS toxicity when busulfan was administered according to the body surface area. More pharmacodynamic studies are required to establish the relation between the systemic exposure to busulfan and the therapeutic efficacy, especially in young children undergoing BMT or HSCT. In the present time an accurate and effective busulfan plasma level monitoring combined with dose adjustment based on the known pharmacological parameters may improve the clinical outcome for patients undergoing BMT.
Product Description
Supelco
Busulfan, analytical standard, for drug analysis
Busulfan, European Pharmacopoeia (EP) Reference Standard