Merck
  • Impact of mass azithromycin distribution on malaria parasitemia during the low-transmission season in Niger: a cluster-randomized trial.
Impact of mass azithromycin distribution on malaria parasitemia during the low-transmission season in Niger: a cluster-randomized trial.
The American journal of tropical medicine and hygiene (2014-03-13)
Bruce D Gaynor, Abdou Amza, Boubacar Kadri, Baido Nassirou, Ousmane Lawan, Laouali Maman, Nicole E Stoller, Sun N Yu, Stephanie A Chin, Sheila K West, Robin L Bailey, Philip J Rosenthal, Jeremy D Keenan, Travis C Porco, Thomas M Lietman
ABSTRACT
We assessed the effect of mass azithromycin treatment on malaria parasitemia in a trachoma trial in Niger. Twenty-four study communities received treatment during the wet, high-transmission season. Twelve of the 24 communities were randomized to receive an additional treatment during the dry, low-transmission season. Outcome measurements were conducted at the community-level in children < 1-72 months of age in May-June 2011. Parasitemia was higher in the 12 once-treated communities (29.8%, 95% confidence interval [CI] = 21.5-40.0%) than in the 12 twice-treated communities (19.5%, 95% CI = 13.0-26.5%, P = 0.03). Parasite density was higher in once-treated communities (354 parasites/μL, 95% CI = 117-528 parasites/μL) than in twice-treated communities (74 parasites/μL, 95% CI = 41-202 parasites/μL, P = 0.03). Mass distribution of azithromycin reduced malaria parasitemia 4-5 months after the intervention. The results suggest that drugs with antimalaria activity can have long-lasting impacts on malaria during periods of low transmission.
Product Description
Supelco
Azithromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Azithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
Azithromycin, European Pharmacopoeia (EP) Reference Standard
Azithromycin for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Azithromycin Identity, United States Pharmacopeia (USP) Reference Standard
USP
Azithromycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Azithromycin