Merck
  • Canavalia ensiformis urease, Jaburetox and derived peptides form ion channels in planar lipid bilayers.
Canavalia ensiformis urease, Jaburetox and derived peptides form ion channels in planar lipid bilayers.
Archives of biochemistry and biophysics (2014-03-04)
Angela R Piovesan, Anne H S Martinelli, Rodrigo Ligabue-Braun, Jean-Louis Schwartz, Celia R Carlini
ABSTRACT
Ureases catalyze the hydrolysis of urea into NH3 and CO2. They are synthesized by plants, fungi and bacteria but not by animals. Ureases display biological activities unrelated to their enzymatic activity, i.e., platelet and neutrophil activation, fungus inhibition and insecticidal effect. Urease from Canavalia ensiformis (jack bean) is toxic to several hemipteran and coleopteran insects. Jaburetox is an insecticidal fragment derived from jack bean urease. Among other effects, Jaburetox has been shown to interact with lipid vesicles. In this work, the ion channel activity of C. ensiformis urease, Jaburetox and three deletion mutants of Jaburetox (one lacking the N-terminal region, one lacking the C-terminal region and one missing the central β-hairpin) were tested on planar lipid bilayers. All proteins formed well resolved, highly cation-selective channels exhibiting two conducting states whose conductance ranges were 7-18pS and 32-79pS, respectively. Urease and the N-terminal mutant of Jaburetox were more active at negative potentials, while the channels of the other peptides did not display voltage-dependence. This is the first direct demonstration of the capacity of C. ensiformis urease and Jaburetox to permeabilize membranes through an ion channel-based mechanism, which may be a crucial step of their diverse biological activities, including host defense.
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), powder, white, ~8 U/mg
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), powder, ~1 U/mg
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type III, powder, 15,000-50,000 units/g solid
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type III, glycerol solution, 500-800 units/mL
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type IX, powder, 50,000-100,000 units/g solid
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type C-3, powder, ≥600,000 units/g solid
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean)