Merck
  • Tetrahydroisoquinoline and 1-methyl-tetrahydroisoquinoline as novel endogenous amines in rat brain.
Tetrahydroisoquinoline and 1-methyl-tetrahydroisoquinoline as novel endogenous amines in rat brain.
Biochemical and biophysical research communications (1986-10-15)
M Kohno, S Ohta, M Hirobe
ABSTRACT
This is the first report to identify 1,2,3,4-tetrahydroisoquinoline and 1-methyl-1,2,3,4-tetrahydroisoquinoline as endogenous amines from non-treated rat brain. The detection was performed by coupled gas chromatography - multiple ion detection, using heptafluorobutyric anhydride and pentafluoropropionic anhydride as derivatizing reagents. These amines might be endogenous substances inducing parkinsonism.