Merck
  • Hair lightening in an hemodialysis patient treated by heptaminol (heptamyl).
Hair lightening in an hemodialysis patient treated by heptaminol (heptamyl).
Dermatology online journal (2009-05-20)
Leila Gharibi, Kawtar Zouhair, Benyouness Ramdani, Hakima Benchikhi
ABSTRACT
Hair lightening occurred in a patient treated with heptaminol (heptamyl). This patient had chronic renal failure and had been undergoing hemodialysis since 2003. The hair hypopigmentation reversed after stopping the drug. The mechanism of the hair color modification is not understood.
Product Description
Heptaminol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.