- Presenilins: members of the gamma-secretase quartets, but part-time soloists too.
Presenilins: members of the gamma-secretase quartets, but part-time soloists too.
Physiology (Bethesda, Md.) (2008-08-14)
Tomoko Wakabayashi, Bart De Strooper
PMID18697993
ABSTRACT
The presenilins in combination with other proteins generate different gamma-secretases, which are involved in the regulated intramembrane proteolysis of a variety of proteins. Understanding the specificity and regulation of these proteases will potentially lead to novel therapeutics for Alzheimer's disease and cancer. Presenilins appear also to exert additional functions outside of the gamma-secretase quartets, which needs further investigation.