Merck
  • Phosphodiesterase 9A regulates central cGMP and modulates responses to cholinergic and monoaminergic perturbation in vivo.
Phosphodiesterase 9A regulates central cGMP and modulates responses to cholinergic and monoaminergic perturbation in vivo.
The Journal of pharmacology and experimental therapeutics (2012-02-14)
Robin J Kleiman, Douglas S Chapin, Curt Christoffersen, Jody Freeman, Kari R Fonseca, Kieran F Geoghegan, Sarah Grimwood, Victor Guanowsky, Mihály Hajós, John F Harms, Christopher J Helal, William E Hoffmann, Geralyn P Kocan, Mark J Majchrzak, Dina McGinnis, Stafford McLean, Frank S Menniti, Fredrick Nelson, Robin Roof, Anne W Schmidt, Patricia A Seymour, Diane T Stephenson, Francis David Tingley, Michelle Vanase-Frawley, Patrick R Verhoest, Christopher J Schmidt
ABSTRACT
Cyclic nucleotides are critical regulators of synaptic plasticity and participate in requisite signaling cascades implicated across multiple neurotransmitter systems. Phosphodiesterase 9A (PDE9A) is a high-affinity, cGMP-specific enzyme widely expressed in the rodent central nervous system. In the current study, we observed neuronal staining with antibodies raised against PDE9A protein in human cortex, cerebellum, and subiculum. We have also developed several potent, selective, and brain-penetrant PDE9A inhibitors and used them to probe the function of PDE9A in vivo. Administration of these compounds to animals led to dose-dependent accumulation of cGMP in brain tissue and cerebrospinal fluid, producing a range of biological effects that implied functional significance for PDE9A-regulated cGMP in dopaminergic, cholinergic, and serotonergic neurotransmission and were consistent with the widespread distribution of PDE9A. In vivo effects of PDE9A inhibition included reversal of the respective disruptions of working memory by ketamine, episodic and spatial memory by scopolamine, and auditory gating by amphetamine, as well as potentiation of risperidone-induced improvements in sensorimotor gating and reversal of the stereotypic scratching response to the hallucinogenic 5-hydroxytryptamine 2A agonist mescaline. The results suggested a role for PDE9A in the regulation of monoaminergic circuitry associated with sensory processing and memory. Thus, PDE9A activity regulates neuronal cGMP signaling downstream of multiple neurotransmitter systems, and inhibition of PDE9A may provide therapeutic benefits in psychiatric and neurodegenerative diseases promoted by the dysfunction of these diverse neurotransmitter systems.
Sigma-Aldrich
PF-04449613, ≥98% (HPLC)