Merck
  • Multiresidue analysis of pesticides in fruits and vegetables using solid-phase extraction and gas chromatographic methods.
Multiresidue analysis of pesticides in fruits and vegetables using solid-phase extraction and gas chromatographic methods.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2008-02-14)
Asha Lal, Guanhuat Tan, Meekin Chai
ABSTRACT
A new extraction and cleanup procedure with gas chromatography was developed for the sensitive determination of acephate, dimethoate, malathion, diazinon, quinalphos, chlorpyrifos, profenofos, alpha-endosulfan, beta-endosulfan, chlorothalonil and carbaryl using 1-chloro-4-fluorobenzene as an internal standard in fruits and vegetables. Several extracting and eluting solvents for solid-phase extraction were investigated. The overall extracting solvent with a mixture of acetone:ethyl acetate:hexane (10:80:10, v/v/v) and a eluting solvent of 5% acetone in hexane used with the RPC18 cartridge gave the best recovery for all of the investigated pesticides, and minimized the interference from co-extractants. Under the optimal extraction and clean-up conditions, recoveries of 85 - 99% with RSD < 5.0% (n = 3) for most of the pesticides at the 0.02 - 0.5 mg/kg level were obtained. The limit of detection was between 0.005 - 0.01 mg/kg and the limit of quantification was 0.01 mg/kg. This analytical procedure was characterized with high accuracy and acceptable sensitivity to meet requirements for monitoring pesticides in crops.
Sigma-Aldrich
1-Chloro-4-fluorobenzene, 98%