MilliporeSigma
  • An improved protocol for the preparation of 3-pyridyl- and some arylboronic acids.
An improved protocol for the preparation of 3-pyridyl- and some arylboronic acids.
The Journal of organic chemistry (2002-07-20)
Wenjie Li, Dorian P Nelson, Mark S Jensen, R Scott Hoerrner, Dongwei Cai, Robert D Larsen, Paul J Reider
ABSTRACT
3-Pyridylboronic acid was prepared in high yield and bulk quantity from 3-bromopyridine via a protocol of lithium-halogen exchange and "in situ quench". This technique was further studied and evaluated on other aryl halides in the preparation of arylboronic acids.