Merck
  • Evaluation of the effects of exposure to organic solvents and hazardous noise among US Air Force Reserve personnel.
Evaluation of the effects of exposure to organic solvents and hazardous noise among US Air Force Reserve personnel.
Noise & health (2013-11-16)
Hayley Hughes, Katherine L Hunting
ABSTRACT
Hearing loss affects many workers including those in the military and may be caused by noise, medications, and chemicals. Exposures to some chemicals may lead to an increase in the incidence of hearing loss when combined with hazardous noise. This retrospective study evaluated the risk for hearing loss among Air Force Reserve personnel exposed to occupational noise with and without exposures to toluene, styrene, xylene, benzene, and JP-8 (jet fuel). Risk factors associated with hearing loss were determined using logistic and linear regression. Stratified analysis was used to evaluate potential interaction between solvent and noise exposure. The majority of the subjects were male (94.6%) and 35 years or older on the date of their first study audiogram (66%). Followed for an average of 3.2 years, 9.2% of the study subjects had hearing loss in at least one ear. Increasing age (odds ratio [OR] = 1.03 per year of age) and each year of follow-up time (OR = 1.23) were significantly associated with hearing loss. Low and moderate solvent exposures were not associated with hearing loss. Linear regression demonstrated that hearing loss was significantly associated with age at first study audiogram, length of follow-up time, and exposure to noise. Hearing decreased by 0.04 decibels for every decibel increase in noise level or by almost half a decibel (0.4 dB) for every 10 decibel increase in noise level.
Product Description
Supelco
Styrene, analytical standard
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluene, analytical standard
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Supelco
Benzene, analytical standard
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%