Merck
  • Collective intermolecular motions dominate the picosecond dynamics of short polymer chains.
Collective intermolecular motions dominate the picosecond dynamics of short polymer chains.
Physical review letters (2013-11-12)
Humphrey Morhenn, Sebastian Busch, Hendrik Meyer, Dieter Richter, Winfried Petry, Tobias Unruh
ABSTRACT
Neutron scattering and extensive molecular dynamics simulations of an all atom C(100)H(202) system were performed to address the short-time dynamics leading to center-of-mass self-diffusion. The simulated dynamics are in excellent agreement with resolution resolved time-of-flight quasielastic neutron scattering. The anomalous subdiffusive center-of-mass motion could be modeled by explicitly accounting for viscoelastic hydrodynamic interactions. A model-free analysis of the local reorientations of the molecular backbone revealed three relaxation processes: While two relaxations characterize local bond rotation and global molecular reorientation, the third component on intermediate times could be attributed to transient flowlike motions of atoms on different molecules. The existence of these collective motions, which are clearly visualized in this Letter, strongly contribute to the chain relaxations in molecular liquids.
Product Description
Polyethylene (LDPE), ERM®, certified reference material
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC