Merck
Direct transfer of graphene onto flexible substrates.
Proceedings of the National Academy of Sciences of the United States of America (2013-10-16)
Luiz G P Martins, Yi Song, Tingying Zeng, Mildred S Dresselhaus, Jing Kong, Paulo T Araujo
ABSTRACT
In this paper we explore the direct transfer via lamination of chemical vapor deposition graphene onto different flexible substrates. The transfer method investigated here is fast, simple, and does not require an intermediate transfer membrane, such as polymethylmethacrylate, which needs to be removed afterward. Various substrates of general interest in research and industry were studied in this work, including polytetrafluoroethylene filter membranes, PVC, cellulose nitrate/cellulose acetate filter membranes, polycarbonate, paraffin, polyethylene terephthalate, paper, and cloth. By comparing the properties of these substrates, two critical factors to ensure a successful transfer on bare substrates were identified: the substrate's hydrophobicity and good contact between the substrate and graphene. For substrates that do not satisfy those requirements, polymethylmethacrylate can be used as a surface modifier or glue to ensure successful transfer. Our results can be applied to facilitate current processes and open up directions for applications of chemical vapor deposition graphene on flexible substrates. A broad range of applications can be envisioned, including fabrication of graphene devices for opto/organic electronics, graphene membranes for gas/liquid separation, and ubiquitous electronics with graphene.
Product Description
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 4,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 100,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 10,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 50,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 20,000
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Cellulose acetate, average Mn ~30,000 by GPC
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,480,000
Sigma-Aldrich
Graphite, powder, <20 μm, synthetic
Sigma-Aldrich
Graphite, flakes
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Sigma-Aldrich
Graphite, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis
Sigma-Aldrich
Graphite, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Sigma-Aldrich
Cellulose acetate, average Mn ~50,000 by GPC
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 8,000
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Polytetrafluoroethylene preparation, 60 wt % dispersion in H2O
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm