Merck
Substrate-induced rearrangements in glutamate-transporter homologs.
Nature structural & molecular biology (2013-10-08)
Baruch I Kanner
ABSTRACT
Sodium-coupled glutamate transporters regulate excitatory signaling in the brain. A new crystal structure shows how the substrate induces changes in the binding pocket of an archaeal transporter homolog, providing new insights into the mechanism of transport.
Product Description
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Aspartic acid hemimagnesium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)