Merck
  • Implications of the O-GlcNAc modification in the regulation of nuclear apoptosis in T cells.
Implications of the O-GlcNAc modification in the regulation of nuclear apoptosis in T cells.
Biochimica et biophysica acta (2013-09-17)
Bruno Johnson, Marlyse Opimba, Jacques Bernier
ABSTRACT
O-linked β-N-acetylglucosamine (O-GlcNAc) is a nutrient-/stress-sensitive post-translational modification that affects nucleocytoplasmic proteins. The enzyme O-N-acetylglucosamine transferase (OGT) catalyzes the addition of O-GlcNAc, whereas O-N-acetylglucosaminidase (OGA) removes it. O-GlcNAcylation plays a role in fundamental regulatory mechanisms through the modification of proteins involved in cell division, metabolism, transcription, cell signaling and apoptosis. The effects of O-GlcNAcylation on apoptosis appear to be cell-dependent, as elevated levels played a protective role in primary neonatal rat ventricular myocytes but had a cytotoxic effect in rat pancreatic β-cells. The aim of the current study was to determine the implications of the O-GlcNAc modification on T cell apoptosis. Human T lymphoblastic HPB-ALL cells were treated with the OGA inhibitor O-(2-acetamido-2-deoxy-d-glucopyranosylidene) amino-N-phenylcarbamate (PUGNAc), or with glucosamine (GlcN), to increase O-GlcNAcylation. Apoptosis was induced in the presence of tributyltin (TBT). DNA fragmentation was observed by cell cycle analysis and corresponded to the sub G0/G1 population. O-GlcNAcylated proteins were detected by immunoblot using a specific antibody (ctd110.6) and were precipitated using succinylated wheat germ agglutinin (sWGA). HPB-ALL cells treated with PUGNAc displayed a significant reduction in DNA fragmentation after TBT-induced apoptosis. DFF45, the protein that inhibits the endonuclease DFF40, was identified to be O-GlcNAc modified. O-GlcNAcylated DFF45 appeared to be more resistant to caspase cleavage during apoptosis. Our results suggest that a decrease in the O-GlcNAc modification on DFF45 occurs before its cleavage by caspase. Our results indicate that the O-GlcNAcylation of DFF45 may represent a mechanism to control the accidental activation of DFF.
Sigma-Aldrich
Anti-DFF45/35 Antibody, NT, Chemicon®, from rabbit
Sigma-Aldrich
Anti-β-O-Linked N-Acetylglucosamine antibody, Mouse monoclonal, clone CTD110.6, purified from hybridoma cell culture
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥95% (HPLC)
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥99% (HPLC)
Sigma-Aldrich
N-Acetyl-D-glucosamine, BioReagent, suitable for cell culture
Sigma-Aldrich
PUGNAc, ≥95% (HPLC)
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Tributyltin hydride, contains 0.05% BHT as stabilizer, 97%