Merck
  • Cicer α-galactosidase immobilization onto functionalized graphene nanosheets using response surface method and its applications.
Cicer α-galactosidase immobilization onto functionalized graphene nanosheets using response surface method and its applications.
Food chemistry (2013-09-05)
Neelesh Singh, Garima Srivastava, Mahe Talat, Himanshu Raghubanshi, Onkar Nath Srivastava, Arvind M Kayastha
ABSTRACT
Cicer α-galactosidase was immobilized onto functionalized graphene with immobilization efficiency of 84% using response surface methodology (Box-Behnken design). The immobilized enzyme had higher thermal stability than the soluble one, attractive for industrial applications. Immobilization of the enzyme lowered the Km to 1/3rd compared to the soluble enzyme. Raffinose family oligosaccharides (RFOs) are mainly responsible for flatulence by taking soybean derived food products. The immobilized enzyme can be used effectively for the hydrolysis of RFOs. After ten successive runs, the immobilized enzyme still retained approximately 60% activity, with soybean RFOs. The easy availability of enzyme source, ease of its immobilization on matrices, non-toxicity, increased stability of immobilized enzyme and effective hydrolysis of RFOs increase the Cicer α-galactosidase application in food processing industries.
Sigma-Aldrich
α-Galactosidase from green coffee beans, ammonium sulfate suspension, ≥9 units/mg protein
Sigma-Aldrich
Graphite, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Graphite, powder, <20 μm, synthetic
Sigma-Aldrich
Graphite, flakes
Sigma-Aldrich
Graphite, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis
Sigma-Aldrich
α-Galactosidase, positionally specific from Escherichia coli, recombinant, expressed in E. coli, buffered aqueous solution