Merck
A presynaptic ENaC channel drives homeostatic plasticity.
Neuron (2013-08-27)
Meg A Younger, Martin Müller, Amy Tong, Edward C Pym, Graeme W Davis
ABSTRACT
An electrophysiology-based forward genetic screen has identified two genes, pickpocket11 (ppk11) and pickpocket16 (ppk16), as being necessary for the homeostatic modulation of presynaptic neurotransmitter release at the Drosophila neuromuscular junction (NMJ). Pickpocket genes encode Degenerin/Epithelial Sodium channel subunits (DEG/ENaC). We demonstrate that ppk11 and ppk16 are necessary in presynaptic motoneurons for both the acute induction and long-term maintenance of synaptic homeostasis. We show that ppk11 and ppk16 are cotranscribed as a single mRNA that is upregulated during homeostatic plasticity. Acute pharmacological inhibition of a PPK11- and PPK16-containing channel abolishes the expression of short- and long-term homeostatic plasticity without altering baseline presynaptic neurotransmitter release, indicating remarkable specificity for homeostatic plasticity rather than NMJ development. Finally, presynaptic calcium imaging experiments support a model in which a PPK11- and PPK16-containing DEG/ENaC channel modulates presynaptic membrane voltage and, thereby, controls calcium channel activity to homeostatically regulate neurotransmitter release.
Product Description
USP
Amiloride hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
5-(N-Ethyl-N-isopropyl)amiloride