Merck
  • Styrene N-vinylpyrrolidone metal-nanocomposites as antibacterial coatings against Sulfate Reducing Bacteria.
Styrene N-vinylpyrrolidone metal-nanocomposites as antibacterial coatings against Sulfate Reducing Bacteria.
Materials science & engineering. C, Materials for biological applications (2013-08-06)
M Fathy, A Badawi, A M Mazrouaa, N A Mansour, E A Ghazy, M Z Elsabee
ABSTRACT
Copolymer of styrene, and vinylpyrrolidone was prepared by various techniques. Different nanometals and nanometal oxides were added into the copolymer as antimicrobial agents against Sulfate Reducing Bacteria (SRB). The nanocomposite chemical structure was confirmed by using FTIR, (1)H NMR spectroscopy and thermogravimetric analysis (TGA). The biocidal action of these nanocomposites against the SRB was detected using sulfide determination method in Postgate medium B. The data indicated that the nanocomposites had an inhibitory effect on the growth of SRB and reduced the bacterial corrosion rate of mild steel coupons. The prepared nanocomposites have high inhibition efficiency when applied as coatings and show less efficiency when applied as solids or solution into SRB medium. The copolymer and its nanocomposites effectively reduced the total corrosion rate as determined by total weight loss method.
Product Description
Supelco
Styrene, analytical standard
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Sigma-Aldrich
1-Vinyl-2-pyrrolidinone, contains sodium hydroxide as inhibitor, ≥99%