Merck
  • Detecting phase separation of freeze-dried binary amorphous systems using pair-wise distribution function and multivariate data analysis.
Detecting phase separation of freeze-dried binary amorphous systems using pair-wise distribution function and multivariate data analysis.
International journal of pharmaceutics (2013-07-23)
Norman Chieng, Hjalte Trnka, Johan Boetker, Michael Pikal, Jukka Rantanen, Holger Grohganz
ABSTRACT
The purpose of this study is to investigate the use of multivariate data analysis for powder X-ray diffraction-pair-wise distribution function (PXRD-PDF) data to detect phase separation in freeze-dried binary amorphous systems. Polymer-polymer and polymer-sugar binary systems at various ratios were freeze-dried. All samples were analyzed by PXRD, transformed to PDF and analyzed by principal component analysis (PCA). These results were validated by differential scanning calorimetry (DSC) through characterization of glass transition of the maximally freeze-concentrate solute (Tg'). Analysis of PXRD-PDF data using PCA provides a more clear 'miscible' or 'phase separated' interpretation through the distribution pattern of samples on a score plot presentation compared to residual plot method. In a phase separated system, samples were found to be evenly distributed around the theoretical PDF profile. For systems that were miscible, a clear deviation of samples away from the theoretical PDF profile was observed. Moreover, PCA analysis allows simultaneous analysis of replicate samples. Comparatively, the phase behavior analysis from PXRD-PDF-PCA method was in agreement with the DSC results. Overall, the combined PXRD-PDF-PCA approach improves the clarity of the PXRD-PDF results and can be used as an alternative explorative data analytical tool in detecting phase separation in freeze-dried binary amorphous systems.
Sigma-Aldrich
Kollidon® 25
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
Povidone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Povidone, meets USP testing specifications
Supelco
Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Supelco
Dextran from Leuconostoc mesenteroides, for GPC, analytical standard, Mw 50,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 670,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 5,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 25,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 410,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~60,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Supelco
Poly(vinylpolypyrrolidone), ~110 μm particle size
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran, enzymatic synth.
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)