Merck
  • Silver nanoparticles-decorated polyphosphazene nanotubes: synthesis and applications.
Silver nanoparticles-decorated polyphosphazene nanotubes: synthesis and applications.
Nanoscale (2013-07-16)
Minghuan Wang, Jianwei Fu, Dandan Huang, Chao Zhang, Qun Xu
ABSTRACT
Herein, we report the preparation of poly (cyclotriphosphazene-co-4,4'-sulfonyldiphenol) (PZS) nanotubes decorated with Ag nanoparticles (NPs). The PZS nanotubes have been synthesized firstly via an in situ template approach, and then Ag nanoparticles were prepared via in situ reduction of AgNO3 with NaBH4 as the reductant, supported on the surface of the PZS nanotubes. The as-obtained PZS nanotubes and PZS@Ag NPs composites were characterized by means of Fourier transform infrared spectroscopy (FT-IR), scanning electron microscopy (SEM), transmission electron microscopy (TEM) equipped with energy-dispersive X-ray spectroscopy (EDS), X-ray diffraction (XRD), atomic absorption spectroscopy (AAS), thermogravimetric analysis (TGA), and UV-vis spectroscopy. Characterization results showed that Ag NPs with good dispersibility were well anchored onto the surface of the PZS nanotubes with superior thermal stability. In addition, the catalytic activities and reusability of these composites were investigated by employing the reduction of 4-nitrophenol (4-NP) into 4-aminophenol (4-AP) by NaBH4 as a model reaction.
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
VenPure® SF, powder
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Supelco
4-Aminophenol (Acetaminophen Related Compound K) (Paracetamol Impurity K), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Supelco
4-Aminophenol, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
4-Aminophenol, ≥98%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis