Merck
  • Enhanced production of pigments by addition of surfactants in submerged fermentation of Monascus purpureus H1102.
Enhanced production of pigments by addition of surfactants in submerged fermentation of Monascus purpureus H1102.
Journal of the science of food and agriculture (2013-04-19)
Yonghui Wang, Bobo Zhang, Liping Lu, Yan Huang, Ganrong Xu
ABSTRACT
The production of pigments by Monascus spp. has attracted increasing attention. Modification of the cell membrane structure by addition of surfactants has proved to be effective for the secretion of intracellular metabolites. Hence in this study the effects and underlying mechanism of surfactants on the production of pigments in submerged fermentation of Monascus purpureus H1102 were systematically investigated. Various surfactants exerted significant but different impacts on the biomass and production of pigments. The maximum production of pigment (304.3 U mL(-1) ) and highest extracellular/intracellular pigment ratio (1.46) were achieved when 15 g L(-1) Triton X-100 was added at 24 h of fermentation, corresponding to significant increases of 88.4 and 240% respectively compared with the control. Meanwhile, the concentration of citrinin (0.94 mg L(-1) ) was 20.6% lower than that of the control. A further study on the fatty acid composition of M. purpureus H1102 showed that the unsaturated/saturated fatty acid ratio and the index of unsaturated fatty acid increased significantly with the addition of Triton X-100. The addition of surfactant Triton X-100 could greatly enhance the production of pigment. It was suggested that Triton X-100 facilitated the secretion of intracellular pigment and therefore enhanced pigment production accordingly.
Sigma-Aldrich
Triton X-100
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Triton X-45
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-100, peroxide- and carbonyl-free
Supelco
Citrinin solution, ~100 μg/mL in acetonitrile, analytical standard
Octoxinol 10, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Triton X-102
Sigma-Aldrich
Triton X-305 solution, 70% in H2O
Sigma-Aldrich
Citrinin, from Penicillium citrinum, ≥98% (HPLC)
Sigma-Aldrich
Triton X-100 solution, BioUltra, ~10% in H2O