Merck
  • Alpha-tocopherol in the ventricular cerebrospinal fluid of Parkinson's disease patients: dose-response study and correlations with plasma levels.
Alpha-tocopherol in the ventricular cerebrospinal fluid of Parkinson's disease patients: dose-response study and correlations with plasma levels.
Neurology (1996-10-01)
E J Pappert, C C Tangney, C G Goetz, Z D Ling, J W Lipton, G T Stebbins, P M Carvey
ABSTRACT
To determine if ventricular cerebrospinal fluid (vCSF) alpha-tocopherol levels in Parkinson's disease (PD) patients can be increased by oral alpha-tocopherol supplementation and whether vCSF levels are linearly related to plasma alpha-tocopherol levels. In spite of its putative neuroprotective properties, alpha-tocopherol has failed to alter PD clinical progression. However, the ability of supplemental alpha-tocopherol to affect brain or vCSF levels has never been assessed in humans nor has a dose response curve for alpha-tocopherol in vCSF been established. Five PD patients with Ommaya catheters received oral dl-alpha-tocopherol over 5 months. Each patient ingested alpha-tocopherol daily with monthly dosage increases (400, 800, 1,600, 3,200, 4,000 IU/day). Plasma and vCSF samples were obtained at baseline and at the end of each month. Alpha-tocopherol levels were determined in triplicate by high-pressure liquid chromatography with fluorometric and electrochemical detection. At baseline, endogenous alpha-tocopherol was detected in plasma and vCSF, with a greater than one-hundred-fold difference between the fluid compartments (mean plasma level 18.76 microM/l (SD +/- 4.69) versus mean CSF level 0.114 microM/l (SD +/- 0.084). A clear dose-response curve occurred in plasma, with statistically significant increases over baseline developing even with 400 IU/d. With higher doses, a significant increase continued without evidence of saturation. However, there was no significant increase in vCSF alpha-tocopherol levels at any dose, including the supraclinical (4,000 IU/d). There was no correlation between plasma and vCSF alpha-tocopherol levels. Oral alpha-tocopherol supplementation, even at supraclinical doses, fails to increase vCSF alpha-tocopherol levels. This lack of change may be due to limited passage across the blood-brain barrier or very rapid alpha-tocopherol metabolism. All prior negative studies on efficacy of alpha-tocopherol in PD may need reevaluation in light of these pharmacologic data.
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Supelco
L-Cystine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
L-Cystine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.5%
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Strontium, granular, 99% trace metals basis
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular