Merck
  • High-performance liquid chromatographic enantioseparation of beta-amino acids.
High-performance liquid chromatographic enantioseparation of beta-amino acids.
Journal of chromatography. A (2001-09-15)
A Péter, L Lázár, F Fülöp, D W Armstron
ABSTRACT
Direct and indirect high-performance liquid chromatographic methods were developed for the enantioseparation of beta-amino acids (beta-substituted-beta-alanines). Direct separation involved the application of chiral columns: Crownpak CR(+), Chirobiotic T and Chirobiotic R. Indirect separation was based on precolumn derivatization with 2,3,4,6-tetra-O-acetyl-beta-D-glucopyranosyl isothiocyanate or N-alpha-(2,4-dinitro-5-fluorophenyl)-L-alanineamide (Marfey's reagent), with subsequent separation on an achiral column. The chromatographic conditions were varied to achieve optimum separation.
Product Description
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm