Merck
  • A circular dichroism and fluorescence quenching study of the interactions between rhodium(II) complexes and human serum albumin.
A circular dichroism and fluorescence quenching study of the interactions between rhodium(II) complexes and human serum albumin.
Journal of inorganic biochemistry (1999-07-14)
B P Espósito, A Faljoni-Alário, J F de Menezes, H F de Brito, R Najjar
ABSTRACT
Various divalent rhodium complexes Rh2(L)4 (L = acetate, propionate, butyrate, trifluoroacetate and trifluoroacetamidate) have been found to bind to non-defatted human serum albumin (HSA) at molar ratios about 8:1. The circular dichroism measurements showed that the more liposoluble carboxylates, butyrate and trifluoroacetate, caused the major alterations of the secondary structure of HSA. Stern-Volmer constants for the fluorescence quenching of the buried Trp214 residue by these complexes were also higher for the lipophilic metal compounds. In the case of the rhodium carboxylates it was observed that their denaturating and quenching properties could be explained in terms of their liposolubilities: the higher their lipophilic characters, the higher their abilities to penetrate inside the protein framework leading to structural alterations, and the closer they could get to the Trp residue causing fluorescence quenching. The liposoluble amidate complex, Rh2 (tfc)4, presented an intermediate quenching and did not cause structural alterations in the protein, presumably not penetrating inside the peptidic backbone. This study shows that it is possible to design new antitumor metal complexes which bind, to a large extent, to a transport protein causing little structural damage.
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Supelco
Propionic acid, analytical standard
Sigma-Aldrich
Propionic acid, ≥99.5%