Merck
  • Adenosine A1 receptor-dependent and -independent effects of the allosteric enhancer PD 81,723.
Adenosine A1 receptor-dependent and -independent effects of the allosteric enhancer PD 81,723.
The Journal of pharmacology and experimental therapeutics (1999-01-26)
B Musser, R V Mudumbi, J Liu, R D Olson, R E Vestal
ABSTRACT
The 2-amino-3-benzoylthiophene PD 81,723 has been shown to exhibit allosteric enhancement of adenosine A1 receptor binding and function. The aim of this study was to clarify the mechanism of this effect using membranes purified from rat brain and Chinese hamster ovary (CHO)-A1 cells that stably express the rat adenosine A1 receptor as well as intact CHO-A1 and nontransfected CHO cells. In membranes containing 100 microM magnesium, (2-amino-4, 5-dimethyl-3-thienyl)-[3-(trifluoromethyl)phenyl]methanone (PD 81, 723) significantly increased the affinity of the adenosine A1 receptor agonist, cyclopentyladenosine, for the low-affinity receptor without affecting high-affinity binding or Bmax. In intact cells, PD 81,723 inhibited basal adenylyl cyclase (AC) activity as well as forskolin-, cholera toxin-, and pertussis toxin-stimulated AC activity in CHO-A1 and CHO cells. Basal AC activity was inhibited 49% in CHO and 82% in CHO-A1 cells by 30 microM PD 81,723. In CHO-A1 cells, half-maximal inhibition of forskolin-stimulated AC occurred at 5 microM PD 81,723 compared to 10 microM in CHO cells. Cholera toxin-stimulated AC was reduced 90% in both CHO and CHO-A1 cells by 30 microM PD 81,723. At the same concentration of PD 81,723, pertussis toxin-stimulated AC activity was reduced 86% (CHO-A1) and 77% (CHO). [3H]forskolin was displaced from purified rat liver AC by PD 81,723 with an IC50 of 96 microM. These results demonstrate that two mechanisms appear to contribute to the observed effects of PD 81, 723. One mechanism is allosteric enhancement of adenosine A1 receptor function. Results from transfected and nontransfected cells suggest that PD 81,723 also inhibits AC directly by binding to the catalytic unit at or near the forskolin-binding site.