Merck
  • Prolyl hydroxylase inhibitor and lysyl hydroxylase inhibitor inhibit spreading of corneal epithelium.
Prolyl hydroxylase inhibitor and lysyl hydroxylase inhibitor inhibit spreading of corneal epithelium.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie (1994-08-01)
S Saika, N Hashizume, Y Okada, S Kobata, O Yamanaka, K Uenoyama, A Ooshima
ABSTRACT
The spreading of epithelium is critical in the healing of corneal wounds. Such epithelial spreading requires the continuous production of protein and glycoprotein. To determine whether collagen production is required for the spreading of corneal epithelium, we studied the effects of inhibitors for collagen production on spreading of corneal epithelium in vitro. We examined the effect of two proline analogs, L-azetidine 2-carboxylic acid and cis-hydroxyproline, a prolyl hydroxylase inhibitor, ethyl-3,4-dihydroxybenzoate, and a lysyl hydroxylase inhibitor, minoxidil, on the spreading of epithelium of organ-cultured rabbit cornea. Both analogs and inhibitors inhibited epithelial spreading in a dose-dependent manner. These observations indicate that collagen production may be involved in the spreading of corneal epithelium.
Sigma-Aldrich
Protocatechuic acid ethyl ester, 97%