Merck
Variation of the alkyl side chain in delta 8-THC.
Life sciences (1995-01-01)
J W Huffman, J A Lainton, D Dai, R D Jordan, S G Duncan
ABSTRACT
The synthesis of (2'RS)-2'-methyl-, (3'RS)-, (3'S)-3'-methyl-, and 4'-methyl-delta 8-THC has been carried out, and the pharmacology of all four compounds has been investigated. All four compounds showed typical cannabinoid activity both in vitro and in vivo. The 2'-methyl compound is somewhat more active than delta 8-THC, while the 4'-methyl isomer is less active. The 3'-methyl-delta 8-THC has approximately the same activity as the parent cannabinoid.
Product Description
Supelco
(−)-Δ8-THC solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®