- The determination of propiopromazine in animal tissue.
The determination of propiopromazine in animal tissue.
Journal of veterinary pharmacology and therapeutics (1981-12-01)
M Olling, R W Stephany, A G Rauws
PMID7349344
ABSTRACT
A thin-layer chromatographic method for the determination of the veterinary tranquillizer propiopromazine (Combelen) in slaughter pigs is described. Propiopromazine residue depletion from the kidney occurs with different rates in sows and in castrated boars. In the liver residues increase over at least 24 h after intramuscular administration, in accordance with the distribution pattern of phenothiazine tranquilizers.