Merck
  • The isolation of human-erythrocyte band-3 polypeptide labelled with a photosensitive hydrophobic probe.
The isolation of human-erythrocyte band-3 polypeptide labelled with a photosensitive hydrophobic probe.
The Biochemical journal (1980-06-01)
E Wells, J B Findlay
ABSTRACT
To investigate the intramembranous domains of the major band-3 polypeptide, human erythrocyte membranes were labelled with 1-azido-4-[125I]iodobenzene. The anion-exchange protein has been isolated by a new procedure that decreases possible contamination by other integral membrane proteins of similar molecular weight. After chemical modification, the protein was subjected to proteolytic cleavage by several proteinases to yield a distinct pattern of peptides, a few of which contained radioactivity.