Merck
Neuroleptic malignant syndrome.
Southern medical journal (1986-03-01)
M P Rampertaap
ABSTRACT
The neuroleptic malignant syndrome (NMS) is a rare but potentially lethal disorder associated with the administration of neuroleptic agents. This syndrome may be underdiagnosed because it is poorly understood and often unrecognized. It affects all age groups and has a 20% mortality. Presenting features include extrapyramidal symptoms, altered mental consciousness, autonomic dysfunction, and hyperthermia. The underlying explanation for these manifestations is a disturbance of the dopaminergic system within the basal ganglia and hypothalamus. Dantrolene (Dantrium), amantadine (Symmetrel), and bromocriptine mesylate (Parlodel) have been efficacious in conjunction with supportive therapy. I report three cases successfully treated with bromocriptine and supportive therapy.
Product Description
Fluphenazine enantate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.