Merck
  • Liquid chromatographic assay and pharmacokinetics of quazepam and its metabolites following sublingual administration of quazepam.
Liquid chromatographic assay and pharmacokinetics of quazepam and its metabolites following sublingual administration of quazepam.
Pharmaceutical research (1988-06-01)
S K Gupta, E H Ellinwood
ABSTRACT
A reverse-phase liquid chromatographic method is described for simultaneous quantification of quazepam, and two of its metabolites, 2-oxoquazepam and N-desaklyl-2-oxoquazepam. The method uses a solid-phase extraction procedure to prepare plasma samples. After extraction, the methanolic extract is evaporated; the residue is then reconstituted in a small volume of mobile phase and chromatographed. The total chromatography time for a single sample is about 20 min. A sensitivity of 1 ng/ml for quazepam and its metabolites is attained when 1 ml of plasma is extracted. Analytical recovery of quazepam and its metabolites added to plasma ranged from 87 to 96%. The maximum within-day and day-to-day coefficients of variation for each compound at concentrations of 20 and 60 ng/ml were 7.6 and 11.2%, respectively. The method was applied to sublingual pharmacokinetic studies of quazepam in healthy volunteers.
Product Description
Supelco
Desalkylflurazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®