Merck
  • Isolation, identification and bacterial mutagenicity of 2-nitro-9-fluorenone from diesel-exhaust particle extracts.
Isolation, identification and bacterial mutagenicity of 2-nitro-9-fluorenone from diesel-exhaust particle extracts.
Mutation research (1986-02-01)
W E Bechtold, T R Henderson, A L Brooks
ABSTRACT
Organic extracts of diesel-exhaust particles show direct mutagenic activity in the Salmonella typhimurium bacterial mutagenicity assay. Nitro-aromatic compounds are believed to be responsible for part of the mutagenicity. A previously unidentified polyfunctional nitro-aromatic compound, 2-nitro-9-fluorenone (2N-Fone) was isolated from diesel-exhaust particles using a two-step fractionation scheme consisting of Sephadex LH20 chromatography and silica-gel thin-layer chromatography. Positive identification was by gas chromatography/mass spectroscopy and coelution with an authentic standard. Direct and indirect mutagenicities of 2N-Fone in several bacterial strains were also determined. The results indicated that 2N-Fone produces 60-70 rev/nmole of direct mutagenic activity, and is about 1/5 to 1/10 as mutagenic as 1-nitropyrene.
Sigma-Aldrich
2-Nitro-9-fluorenone, 99%