Merck
  • Vitamin C intake and breast cancer mortality in a cohort of Swedish women.
Vitamin C intake and breast cancer mortality in a cohort of Swedish women.
British journal of cancer (2013-06-06)
H R Harris, L Bergkvist, A Wolk
ABSTRACT
Vitamin C may influence cancer progression through its antioxidant properties. However, the evidence from observational epidemiologic studies on vitamin C intake and survival following breast cancer diagnosis is not consistent, and the safety of vitamin C supplements following breast cancer diagnosis has not been extensively studied. Using a food-frequency questionnaire we investigated whether vitamin C intake was associated with survival among 3405 women diagnosed with invasive breast cancer in the Swedish Mammography Cohort. From 1987-2010, there were 1055 total deaths with 416 deaths from breast cancer. Women in the highest quartile of pre-diagnosis vitamin C intake had an adjusted HR (95% CI) of breast cancer death of 0.75 (0.57-0.99) compared with those in the lowest quartile (Ptrend=0.03). There was a borderline significant association between vitamin C intake and total mortality (HR=0.84; 95% CI=0.71-1.00; Ptrend=0.08). Among 717 breast cancer cases for whom post-diagnosis supplement use was available, there was no association between vitamin C supplement use (≈1000 mg) and breast cancer-specific mortality (HR=1.06; 95% CI=0.52-2.17). Our findings suggest that dietary vitamin C intake before breast cancer diagnosis may be associated with breast cancer survival. In addition, post-diagnosis vitamin C supplementation at the level observed in our population was not associated with survival.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Pricing and availability is not currently available.
Supelco
L-Ascorbic acid, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Pricing and availability is not currently available.
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Pricing and availability is not currently available.
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Pricing and availability is not currently available.
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.