Merck
  • Nanocrystalline cellulose extraction process and utilization of the byproduct for biofuels production.
Nanocrystalline cellulose extraction process and utilization of the byproduct for biofuels production.
Carbohydrate polymers (2013-03-08)
Sanaa Pirani, Raed Hashaikeh
ABSTRACT
Cellulose consists of amorphous and crystalline regions. It is the crystalline regions which may be exploited to produce nanocrystalline cellulose (NCC). In order to extract nanocrystalline cellulose from native cellulose, sulfuric acid hydrolysis is typically used. The amorphous regions of cellulose are hydrolyzed and degraded into soluble products while the crystalline regions remain intact. In an effort to make the NCC extraction process more feasible, a new process was developed to recover and utilize the hydrolyzed regions of cellulose as a byproduct. The acid hydrolyzed amorphous regions were separated and then recovered (regenerated) into solid particles. XRD data revealed that the recovered material is characteristic of cellulose II. Hydrolysis conditions were optimized to maximize the yield of the recovered material and at the same time produce NCC material. Preliminary experiments showed yield values of approximately 61% for the cellulose I crystalline portions and values of about 21.7% for the recovered material (cellulose II). Enzymatic hydrolysis experiments of the recovered material revealed high susceptibility to enzymatic hydrolysis which makes it a promising source for biofuels production.
Sigma-Aldrich
Sulfuric acid, 99.999%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC