- Structure of (R)-tetrahydro-2-furancarboxylic acid monohydrate.
Structure of (R)-tetrahydro-2-furancarboxylic acid monohydrate.
Acta crystallographica. Section C, Crystal structure communications (1990-01-15)
S Bando, T Takano, R Tanaka, H Iwata, M Ishiguro
PMID2331334
ABSTRACT
C5H10O4, Mr = 134.31, monoclinic, P2(1), a = 11.670 (2), b = 6.177 (3), c = 10.704 (2) A, beta = 115.11 (2) degrees, V = 698.7 (4) A3, Z = 4, Dm = 1.26, Dx = 1.28 g cm-3, Cu K alpha, lambda = 1.54178 A, mu = 9.69 cm-1, F(000) = 288, room temperature, R = 0.046 for 1047 reflections with [Fo[ greater than 3 sigma(F). There are two independent molecules of the title compound in the crystal. No twofold symmetry is observed in the tetrahydrofuran rings. Hydrogen bonds between water molecules and carboxylic acid groups stabilize the crystal structure.